R&D Systems
Manufacturer Code:MAB8400SP
Catalog # MAB8400SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 910825 |
Immunogen | NS0 mouse myeloma cell line transfected with human NRAMP1/SLC11A1 Met1-Gly550 Accession # NP_000569 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Natural resistance-associated macrophage protein 1; natural resistance-associated macrophage protein 1 LSH member 1 NRAMP 1 NRAMP1solute carrier family 11 (sodium/phosphate symporters) member 1 NRAMPnatural resistance-associated macrophage protein 1 SLC11A1 solute carrier family 11 (proton-coupled divalent metal ion transporters); NRAMP 1; solute carrier family 11 (proton-coupled divalent metal ion transporter), member 1; solute carrier family 11 (proton-coupled divalent metal ion transporters), member 1; solute carrier family 11 (sodium/phosphate symporters), member 1; Solute carrier family 11 member 1
Gene Aliases: LSH; NRAMP; NRAMP1; SLC11A1
UniProt ID: (Human) P49279
Entrez Gene ID: (Human) 6556
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.