Invitrogen
Manufacturer Code:PA5110041
Catalog # PIPA5110041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 206-495 of human NR5A2 (NP_003813.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1-fetoprotein transcription factor; Alpha-1-fetoprotein transcription factor AU020803 B1-binding factor B1F B1F2 BOS_16202 CPF CYP7A promoter-binding factor D1Ertd308e FTF FTF/LRH-1 FTZ F1 FTZ F1beta FTZ-F1 FTZ-F1 beta FTZ-F1beta hB1F hB1F 2 hB1F-2 Hepatocytic transcription factor I79_021747 live Liver receptor homolog 1 Lrh1 LRH-1 Nr5a2 Nuclear receptor subfamily 5 group A member 2 OR2.0 SF-2 UF2-H3B; B1-binding factor; b1-binding factor, hepatocyte transcription factor which activates enhancer II of hepatitis B virus; CYP7A promoter-binding factor; fetoprotein transcription factor; fetoprotein-alpha 1 (AFP) transcription factor; FTZ-F1 beta; FTZ-F1 beta2 protein; hB1F; Hepatocytic transcription factor; hepatocytic transcription factor hB1F-3; liver nuclear receptor homolog-1 variant 2; Liver receptor homolog 1; LRH-1; Nuclear receptor subfamily 5 group A member 2; nuclear receptor subfamily 5, group A, member 2
Gene Aliases: AU020803; B1F; B1F2; CPF; D1Ertd308e; FTF; FTZ-F1; FTZ-F1beta; hB1F-2; LRH-1; LRH1; NR5A2; UF2-H3B
UniProt ID: (Human) O00482, (Rat) Q9QWM1, (Mouse) P45448
Entrez Gene ID: (Human) 2494, (Rat) 60349, (Mouse) 26424
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.