Abnova
Manufacturer Code:PAB27773
Catalog # 89-129-292
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 16 amino acid at ligand-binding domain of human NR5A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of NR5A1.
For Research Use Only
Protein Aliases: AD4BP; adrenal 4 binding protein; Adrenal 4-binding protein; ELP; FTZ1; FTZF1; Fushi tarazu factor homolog 1; nuclear receptor AdBP4; Nuclear receptor subfamily 5 group A member 1; nuclear receptor subfamily 5, group A, member 1; SF-1; SF1; Steroid hormone receptor Ad4BP; Steroidogenic factor 1; steroidogenic factor 1 nuclear receptor; steroidogenic factor-1; STF-1
Gene Aliases: AD4BP; ELP; FTZ1; FTZF1; hSF-1; NR5A1; POF7; SF-1; SF1; SPGF8; SRXY3
UniProt ID: (Human) Q13285
Entrez Gene ID: (Human) 2516
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.