Abnova
Manufacturer Code:H00004929M06
Catalog # 89-123-252
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3E10 |
Immunogen | NR4A2 (AAH66890 71 a.a. ~ 170 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant NR4A2.
For Research Use Only
Protein Aliases: HZF-3; Immediate-early response protein NOT; intermediate-early receptor protein; NGFI-B/nur77 beta-type transcription factor homolog; NOT; nuclear receptor related 1; Nuclear receptor subfamily 4 group A member 2; nuclear receptor subfamily 4, group A, member 2; nur related protein-1, human homolog of; NURR1; orphan nuclear receptor NR4A2; Orphan nuclear receptor NURR1; RNR1; T-cell nuclear receptor NOT; TINUR; transcriptionally inducible nuclear receptor related 1; Transcriptionally-inducible nuclear receptor
Gene Aliases: HZF-3; NOT; NR4A2; NURR1; RNR1; TINUR
UniProt ID: (Human) P43354
Entrez Gene ID: (Human) 4929
Molecular Function:
nuclear hormone receptor
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.