Invitrogen
Manufacturer Code:PA532949
Catalog # PIPA532949
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 17 amino acid peptide from internal region of human NUR77. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Early response protein NAK1; GFRP1 HMR N10 NAK-1 NGFIB NP10 NUR77 TR3 ST-59 TR3 orphan receptor early response protein NAK1 growth factor-inducible nuclear protein N10 hormone receptor nerve growth factor IB nuclear receptor variant 1 nuclear hormone receptor NUR/77 nuclear receptor subfamily 4 group A member 1 orphan nucle; growth factor-inducible nuclear protein N10; hormone receptor; nerve growth factor IB nuclear receptor variant 1; Nuclear hormone receptor NUR/77; Nuclear receptor subfamily 4 group A member 1; nuclear receptor subfamily 4, group A, member 1; Nur77; Orphan nuclear receptor HMR; Orphan nuclear receptor TR3; ST-59; steroid receptor TR3; Testicular receptor 3; TR3 orphan receptor
Gene Aliases: GFRP1; HMR; N10; NAK-1; NAK1; NGFIB; NP10; NR4A1; NUR77; TR3
UniProt ID: (Human) P22736
Entrez Gene ID: (Human) 3164
Molecular Function: nuclear hormone receptor receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.