Abnova
Manufacturer Code:PAB27807
Catalog # 89-129-327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 15 amino acid at N-terminus of human NR4A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of NR4A1.
For Research Use Only
Protein Aliases: Early response protein NAK1; GFRP1; growth factor-inducible nuclear protein N10; HMR; hormone receptor; MGC9485; N10; NAK-1; nerve growth factor IB nuclear receptor variant 1; NGFIB; NP10; Nuclear hormone receptor NUR/77; Nuclear receptor subfamily 4 group A member 1; nuclear receptor subfamily 4, group A, member 1; NUR77; Orphan nuclear receptor HMR; Orphan nuclear receptor TR3; ST-59; steroid receptor TR3; Testicular receptor 3; TR3; TR3 orphan receptor
Gene Aliases: GFRP1; HMR; N10; NAK-1; NAK1; NGFIB; NP10; NR4A1; NUR77; TR3
UniProt ID: (Human) P22736
Entrez Gene ID: (Human) 3164
Molecular Function: nuclear hormone receptor receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.