Invitrogen
Manufacturer Code:PA595019
Catalog # PIPA595019
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human NR3C2 identical to the related mouse and rat sequences. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aldosterone receptor; Mineralocorticoid receptor; Mineralocorticoid receptor (aldosterone receptor); mineralocorticoid receptor 1; mineralocorticoid receptor 2; mineralocorticoid receptor delta; Mineralocorticoid receptor MR Nuclear receptor subfamily 3 group C member 2 NR3C2 MCR MLR; MR; Nuclear receptor subfamily 3 group C member 2; nuclear receptor subfamily 3, group C, member 2 variant 3
Gene Aliases: MCR; MLR; MR; NR3C2; NR3C2VIT
UniProt ID: (Human) P08235, (Rat) P22199
Entrez Gene ID: (Human) 4306, (Mouse) 110784, (Rat) 25672
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.