Proteintech
Manufacturer Code:601172IG150UL
Catalog # 50-173-6248
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2H2B8 |
Immunogen | NR2F6 Fusion Protein Ag7614 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EAR 2 EAR2 ERBAL2 NR2F6 V erbA related protein 2; EAR-2; ERBA-related gene-2; Nuclear receptor subfamily 2 group F member 6; nuclear receptor subfamily 2, group F, member 6; nuclear receptor V-erbA-related; v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 2; V-erbA-related protein 2
Gene Aliases: EAR-2; EAR2; ERBAL2; NR2F6
UniProt ID: (Human) P10588
Entrez Gene ID: (Human) 2063
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.