Invitrogen
Manufacturer Code:PA599588
Catalog # PIPA599588
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ARP-1 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2700033K02Rik 9430015G03Rik ADP-ribosylation factor related protein 1 Apolipoprotein AI regulatory protein 1 apolipoprotein A-I regulatory protein 1 apolipoprotein regulatory protein 1 Aporp1 ARP1 ARP-1 chicken ovalbumin chicken ovalbumin upstream promoter transcription factor 2 chicken ovalbumin upstream promoter-transcription factor I CHTD4 COUP transcription factor 2 COUP transcription factor II COUPb COUP-TF II COUP-TF2 COUPTFB COUPTFII COUP-TFII EAR3 MGC117452 NF-E3 NR2F1 NR2F2 nuclear receptor subfamily 2 group F member 2 nuclear receptor subfamily 2 group F member 2 ovalbumin upstream promoter beta nuclear receptor SVP40 Tcfcoup2 TFCOUP2; ADP-ribosylation factor related protein 1; Apolipoprotein A-I regulatory protein 1; apolipoprotein AI regulatory protein 1; ARP-1; chicken ovalbumin upstream promoter transcription factor 2; chicken ovalbumin upstream promoter-transcription factor I; COUP transcription factor 2; COUP transcription factor II; COUP-TF II; COUP-TF2; Nuclear receptor subfamily 2 group F member 2; nuclear receptor subfamily 2, group F, member 2
Gene Aliases: ARP1; CHTD4; COUPTFB; COUPTFII; NF-E3; NR2F1; NR2F2; SVP40; TFCOUP2
UniProt ID: (Human) P24468
Entrez Gene ID: (Human) 7026
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.