Invitrogen
Manufacturer Code:PA546850
Catalog # PIPA546850
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human NR2F2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosylation factor related protein 1; Apolipoprotein A-I regulatory protein 1; apolipoprotein A1 regulatory protein 1; Apolipoprotein AI regulatory protein 1; apolipoprotein regulatory protein 1; ARP-1; chicken ovalbumin upstream promoter transcription factor 2; chicken ovalbumin upstream promoter-transcription factor I; COUP transcription factor 2; COUP transcription factor II; COUP-TF II; COUP-TF2; COUP-TFII; COUPb; COUPTFbeta; Nuclear receptor subfamily 2 group F member 2; Nuclear receptor subfamily 2 group F member 2 ARP1 COUP-TFII COUPTFB MGC117452 SVP40 TFCOUP2 NF-E3 NR2F1 COUPTFII; nuclear receptor subfamily 2, group F, member 1; nuclear receptor subfamily 2, group F, member 2; Ovalbumin upstream promoter beta nuclear receptor; seven-up related 40; svp-related nuclear receptor/retinoid signaling modulator; transcription factor COUP 2
Gene Aliases: 2700033K02Rik; 9430015G03Rik; Aporp1; ARP-1; ARP1; CHTD4; COUP-TF2; COUP-TFII; COUPTFB; COUPTFII; EAR3; NF-E3; NR2F1; NR2F2; SVP40; svp[40]; Tcfcoup2; TFCOUP2; wu:fc94g08
UniProt ID: (Human) P24468, (Bovine) Q9TTR7, (Rat) O09018, (Mouse) P43135
Entrez Gene ID: (Pig) 100153983, (Rabbit) 100346572, (Human) 7026, (Bovine) 281945, (Sheep) 100286739, (Dog) 479026, (Rat) 113984, (Mouse) 11819, (Zebrafish) 30424
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.