Invitrogen
Manufacturer Code:PA528033
Catalog # PIPA528033
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 123 and 383 of Human NR2F2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosylation factor related protein 1; Apolipoprotein A-I regulatory protein 1; Apolipoprotein AI regulatory protein 1; apolipoprotein regulatory protein 1; ARP-1; ARP1 COUPTFB COUPTFII NF-E3 NR2F1 SVP40 TFCOUP2 ADP-ribosylation factor related protein 1 COUP transcription factor 2 COUP transcription factor II apolipoprotein A-I regulatory protein 1 apolipoprotein AI regulatory protein 1 chicken ovalbumin upstream promoter transcription factor 2 chicken ovalbumin; chicken ovalbumin upstream promoter transcription factor 2; chicken ovalbumin upstream promoter-transcription factor I; COUP transcription factor 2; COUP transcription factor II; COUP-TF II; COUP-TF2; Nuclear receptor subfamily 2 group F member 2; nuclear receptor subfamily 2, group F, member 2
Gene Aliases: 2700033K02Rik; 9430015G03Rik; Aporp1; ARP-1; ARP1; CHTD4; COUP-TF2; COUP-TFII; COUPTFB; COUPTFII; EAR3; NF-E3; NR2F1; NR2F2; SVP40; Tcfcoup2; TFCOUP2
UniProt ID: (Human) P24468, (Mouse) P43135
Entrez Gene ID: (Human) 7026, (Mouse) 11819
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.