OriGene
Manufacturer Code:CF807276
Catalog # 50-166-8188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7E6 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-241 of human NR2C2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Nuclear hormone receptor TR4; Nuclear receptor subfamily 2 group C member 2; nuclear receptor subfamily 2, group C, member 2; Orphan nuclear receptor TAK1; Orphan nuclear receptor TR4; orphan receptor TR4; TAK1 TR4; testicular nuclear receptor 4; Testicular receptor 4
Gene Aliases: NR2C2; TAK1; TR4
UniProt ID: (Human) P49116
Entrez Gene ID: (Human) 7182
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.