Proteintech
Manufacturer Code:209811AP150UL
Catalog # 50-173-3186
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hTAK1 NR2C2 Orphan nuclear receptor TAK1 Orphan nuclear receptor TR4 TAK1 Testicular receptor 4 TR2R1 TR4; Nuclear hormone receptor TR4; Nuclear receptor subfamily 2 group C member 2; nuclear receptor subfamily 2, group C, member 2; nuclear receptor subfamily 2, group H, member 2; Orphan nuclear receptor TAK1; Orphan nuclear receptor TR4; orphan receptor TR4; orphan receptor, TR4; testicular nuclear receptor 4; Testicular receptor 4
Gene Aliases: mKIAA4145; Mtr2r1; NR2C2; TAK1; TR4
UniProt ID: (Human) P49116, (Mouse) P49117
Entrez Gene ID: (Human) 7182, (Mouse) 22026
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.