Proteintech
Manufacturer Code:136581AP150UL
Catalog # 50-172-8914
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NR2C1 Fusion Protein Ag4554 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 80-3 cNDA; early embryonic nuclear receptor; mTR2; nr2c1; NR2C1 Orphan nuclear receptor TR2 Testicular receptor 2 TR2; Nuclear receptor subfamily 2 group C member 1; nuclear receptor subfamily 2, group C isoform; nuclear receptor subfamily 2, group C, member 1; nuclear receptor subfamily 2, group H, member 1; Orphan nuclear receptor TR2; orphan receptor, TR2-11; pregnancy specific beta-1-glycoprotein 4; Testicular receptor 2; TR2 nuclear hormone receptor
Gene Aliases: 4831444H07Rik; 80.3; Eenr; NR2C1; Psg4; TR2; TR2-11
UniProt ID: (Human) P13056, (Mouse) Q505F1, (Rat) Q8VIJ4
Entrez Gene ID: (Human) 7181, (Mouse) 22025, (Rat) 252924
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.