Abnova
Manufacturer Code:PAB5910
Catalog # 89-131-999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of mouse Nr1i3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Nr1i3.
For Research Use Only
Protein Aliases: AA209988; AI551208; CAR; CAR-beta; Care2; Constitutive androstane receptor; constitutively transactivates RAREs; ESTM32; MB67; MGC107281; Nuclear receptor subfamily 1 group I member 3; Orphan nuclear receptor MB67
Gene Aliases: AA209988; AI551208; CAR; CAR-beta; Care2; ESTM32; MB67; Nr1i3
UniProt ID: (Mouse) O35627
Entrez Gene ID: (Mouse) 12355
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.