Proteintech
Manufacturer Code:210421AP150UL
Catalog # 50-173-3223
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NR1I3 Fusion Protein Ag15378 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAR; CAR CAR BETA CAR1 Constitutive active response MB67 NR1I3 Orphan nuclear receptor MB67; Constitutive activator of retinoid response; constitutive active receptor; Constitutive active response; constitutive androstane nuclear receptor variant 2; constitutive androstane nuclear receptor variant 3; constitutive androstane nuclear receptor variant 4; constitutive androstane nuclear receptor variant 5; Constitutive androstane receptor; constitutively transactivates RAREs; nuclear receptor constitutive active receptor; Nuclear receptor subfamily 1 group I member 3; nuclear receptor subfamily 1, group I, member 3; orphan nuclear hormone receptor; Orphan nuclear receptor MB67; strain Fischer nuclear receptor (CAR)
Gene Aliases: AA209988; AI551208; CAR; CAR-beta; CAR1; Care2; ESTM32; MB67; NR1I3
UniProt ID: (Human) Q14994, (Mouse) O35627, (Rat) Q9QUS1
Entrez Gene ID: (Human) 9970, (Mouse) 12355, (Rat) 65035
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.