Abnova
Manufacturer Code:PAB19183
Catalog # 89-120-711
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 14 amino acids near N-terminus of human NR1H2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody rasied against synthetic peptide of NR1H2.
For Research Use Only
Protein Aliases: liver X nuclear receptor beta; Liver X receptor beta; LX receptor beta; LXR-b; LXRB; NER; NER-I; nuclear orphan receptor LXR-beta; Nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; nuclear receptor subfamily 1, group H, member 2; Oxysterols receptor LXR-beta; RIP15; steroid hormone-nuclear receptor NER; Ubiquitously-expressed nuclear receptor; UNR
Gene Aliases: LXR-b; LXRB; NER; NER-I; NR1H2; RIP15; UNR
UniProt ID: (Human) E7EWA6
Entrez Gene ID: (Human) 7376
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.