Invitrogen
Manufacturer Code:MA520772
Catalog # PIMA520772
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F4 |
Immunogen | NR1D1 (NP&lowbar068370 233 a.a. &bsim 322 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EAR-1; EAR1 THRA1 THRAL ear-1 hRev Rev-ErbAalpha V-erbA-related protein 1 nuclear receptor Rev-ErbA-alpha nuclear receptor subfamily 1 group D member 1 rev-erbA-alpha; nuclear receptor Rev-ErbA-alpha; Nuclear receptor subfamily 1 group D member 1; nuclear receptor subfamily 1, group D, member 1; Rev-erbA-alpha; Rev-ErbAalpha; V-erbA-related protein 1
Gene Aliases: ear-1; EAR1; HREV; NR1D1; THRA1; THRAL
UniProt ID: (Human) P20393
Entrez Gene ID: (Human) 9572
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.