Invitrogen
Manufacturer Code:PA529865
Catalog # PIPA529865
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 246 and 558 of Human NR1D1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EAR-1; EAR1 THRA1 THRAL ear-1 hRev Rev-ErbAalpha V-erbA-related protein 1 nuclear receptor Rev-ErbA-alpha nuclear receptor subfamily 1 group D member 1 rev-erbA-alpha; nuclear receptor protein; nuclear receptor Rev-ErbA-alpha; Nuclear receptor subfamily 1 group D member 1; nuclear receptor subfamily 1, group D, member 1; orphan nuclear receptor; rev-erb alpha; rev-erbA alpha; rev-erbA(alpha); Rev-erbA-alpha; Rev-ErbA-alpha protein; Rev-ErbAalpha; REV-ERBalpha; V-erbA-related protein 1
Gene Aliases: A530070C09Rik; ear-1; EAR1; HREV; NR1D1; R75201; THRA1; THRAL
UniProt ID: (Human) P20393, (Mouse) Q3UV55
Entrez Gene ID: (Human) 9572, (Mouse) 217166
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.