OriGene
Manufacturer Code:CF806318
Catalog # 50-166-7787
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6D8 |
Immunogen | Full length human recombit protein of human NR0B2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: nuclear receptor SHP; Nuclear receptor subfamily 0 group B member 2; nuclear receptor subfamily 0, group B, member 2; Orphan nuclear receptor SHP; SHP SHP1; Small heterodimer partner
Gene Aliases: NR0B2; SHP; SHP1
UniProt ID: (Human) Q15466
Entrez Gene ID: (Human) 8431
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.