Invitrogen
Manufacturer Code:PA5102494
Catalog # PIPA5102494
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NR0B2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NR0B2 Nr0b2a nuclear receptor SHP nuclear receptor subfamily 0 group B member 2 nuclear receptor subfamily 0 group B member 2 Orphan nuclear receptor SHP Shp SHP1 SHP-1 Small heterodimer partner small heterodimer partner homologue; nuclear receptor SHP; Nuclear receptor subfamily 0 group B member 2; nuclear receptor subfamily 0, group B, member 2; Orphan nuclear receptor SHP; Small heterodimer partner
Gene Aliases: NR0B2; SHP; SHP-1; SHP1
UniProt ID: (Human) Q15466, (Mouse) Q62227
Entrez Gene ID: (Human) 8431, (Mouse) 23957
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.