Abnova
Manufacturer Code:PAB27775
Catalog # 89-129-294
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 16 amino acid at ligand-binding domain of human NR0B2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of NR0B2.
For Research Use Only
Protein Aliases: FLJ17090; nuclear receptor SHP; Nuclear receptor subfamily 0 group B member 2; nuclear receptor subfamily 0, group B, member 2; Orphan nuclear receptor SHP; SHP; SHP1; Small heterodimer partner
Gene Aliases: NR0B2; SHP; SHP1
UniProt ID: (Human) Q15466
Entrez Gene ID: (Human) 8431
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.