Invitrogen
Manufacturer Code:PA532946
Catalog # PIPA532946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 19 amino acid peptide from ligand-binding domain of human DAX1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AHC AHCH AHX DAX-1 DAX1 DSS GTD HHG NROB1 SRXY2 DSS-AHC critical region on the X chromosome protein 1 nuclear hormone receptor nuclear receptor DAX-1 nuclear receptor DAX1 nuclear receptor subfamily 0 group B member 1; DSS-AHC critical region on the X chromosome protein 1; nuclear hormone receptor; Nuclear receptor DAX-1; Nuclear receptor subfamily 0 group B member 1; nuclear receptor subfamily 0, group B, member 1
Gene Aliases: AHC; AHCH; AHX; DAX-1; DAX1; DSS; GTD; HHG; NR0B1; NROB1; SRXY2
UniProt ID: (Human) P51843
Entrez Gene ID: (Human) 190
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.