OriGene
Manufacturer Code:TA504744
Catalog # 50-167-5363
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3C11 |
Immunogen | Full length human recombit protein of human NQO2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DHQV DIA6 NMOR2 QR2; NAD(P)H dehydrogenase, quinone 2; NAD(P)H menadione oxidoreductase-1, dioxin-inducible-2; NAD(P)H quinone dehydrogenase 2; NRH dehydrogenase [quinone] 2; NRH:quinone oxidoreductase 2; QR2; Quinone reductase 2; ribosyldihydronicotinamide dehydrogenase; Ribosyldihydronicotinamide dehydrogenase [quinone]
Gene Aliases: DHQV; DIA6; NMOR2; NQO2; QR2
UniProt ID: (Human) P16083
Entrez Gene ID: (Dog) 606932, (Human) 4835
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.