Invitrogen
Manufacturer Code:MA525981
Catalog # PIMA525981
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G2 |
Immunogen | Full length human recombit protein of NQO2 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DHQVDIA6EC 1.10.99.2MGC94180NAD(P)H dehydrogenase quinone 2NAD(P)H menadione oxidoreductase 1 dioxin inducible 2NAD(P)H menadione oxidoreductase 2 dioxin inducibleNMOR2NQO 2NQO2NQO2&lowbarHUMANNRH dehydrogenase &lsqbquinone] 2NRH dehydrogenaseNRH:quinone oxidoreductase 2OTTHUMP00000015948OTTHUMP00000015949OTTHUMP00000015953Ox 2Ox2QR2QuinoneQuinone reductase 2Ribosyldihydronicotinamide dehydrogenase &lsqbquinone]Ribosyldihydronicotinamide dehydrogenase; NAD(P)H dehydrogenase, quinone 2; NAD(P)H menadione oxidoreductase-1, dioxin-inducible-2; NAD(P)H quinone dehydrogenase 2; NRH dehydrogenase [quinone] 2; NRH:quinone oxidoreductase 2; QR2; Quinone reductase 2; quinone ribosyldihydronicotinamide dehydrogenase; ribosyldihydronicotinamide dehydrogenase; Ribosyldihydronicotinamide dehydrogenase [quinone]; ribosyldihydronicotinamide dehydrogenase quinone
Gene Aliases: DHQV; DIA6; NMOR2; NQO2; QR2
UniProt ID: (Human) P16083
Entrez Gene ID: (Rhesus monkey) 707675, (Dog) 606932, (Human) 4835
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.