Invitrogen
Manufacturer Code:MA535411
Catalog # PIMA535411
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0846 |
Immunogen | A synthesized peptide derived from human NQO2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: DHQV DIA6 EC 1.10.99.2 MGC94180 NMO2 NMOR2 NQO 2 NQO2 NRH dehydrogenase NRH dehydrogenase [quinone] 2 NRH:quinone oxidoreductase 2 OTTHUMP00000015948 OTTHUMP00000015949 OTTHUMP00000015953 Ox 2 Ox2 QR2 Quinone Quinone reductase 2 ribosyldihydronicotinamide Ribosyldihydronicotinamide dehydrogenase [quinone]; NAD(P)H menadione oxidoreductase 2, dioxin inducible; NRH dehydrogenase [quinone] 2; NRH: quinone oxidoreductase; NRH:quinone oxidoreductase 2; QR2; Quinone reductase 2; ribosyldihydronicotinamide dehydrogenase; Ribosyldihydronicotinamide dehydrogenase [quinone]
Gene Aliases: NMO2; Nmor2; Nqo2; Ox2
UniProt ID: (Rat) Q6AY80, (Mouse) Q9JI75
Entrez Gene ID: (Rat) 291084, (Mouse) 18105
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.