Novus Biologicals
Manufacturer Code:NBP273049C
Catalog # NBP273049C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3G4 |
Immunogen | Full length recombinant protein of human NQO-2 (NP_000895) produced in HEK293T cells. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: dioxin-inducible-2 EC 1.10.99.2 NAD(P)H dehydrogenase quinone 2 NAD(P)H menadione oxidoreductase 2 dioxin-inducible NRH dehydrogenase [quinone] 2 NRH:quinone oxidoreductase 2 Quinone reductase 2 ribosyldihydronicotinamide dehydrogenase ribosyldihydronicotinamide dehydrogenase [quinone]; NAD(P)H dehydrogenase, quinone 2; NAD(P)H menadione oxidoreductase-1, dioxin-inducible-2; NRH dehydrogenase [quinone] 2; NRH:quinone oxidoreductase 2; QR2; Quinone reductase 2; ribosyldihydronicotinamide dehydrogenase; Ribosyldihydronicotinamide dehydrogenase [quinone]
Gene Aliases: DHQV; DIA6; NMOR2; NQO2; QR2
UniProt ID: (Human) P16083
Entrez Gene ID: (Human) 4835
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.