Invitrogen
Manufacturer Code:PA577519
Catalog # PIPA577519
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide SFIRKHRRRYSKKTAC corresponding to amino acid residues 291-306 of rat NPY5R |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neuropeptide Y receptor type 5; Neuropeptide Y5 receptor; NPY-Y5 receptor; NPY5-R; NPY5R; NPYY5-R; Y5 receptor
Gene Aliases: Npy5; NPY5-R; Npy5r; NPYR5; NPYY5-R; Y5R
UniProt ID: (Rat) Q63634, (Mouse) O70342
Entrez Gene ID: (Rat) 25340, (Mouse) 18168
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.