Abnova
Manufacturer Code:PAB26441
Catalog # 89-128-082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 16 amino acids at 3rd cytoplasmic domain of human NPY5R. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of NPY5R.
For Research Use Only
Protein Aliases: Neuropeptide Y receptor type 5; NPY-Y5 receptor; NPY5-R; NPYR5; NPYY5-R; Y5 receptor
Gene Aliases: NPY5-R; NPY5R; NPYR5; NPYY5-R
UniProt ID: (Human) Q15761
Entrez Gene ID: (Human) 4889
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.