R&D Systems
Manufacturer Code:FAB10211G100UG
Catalog # FAB10211G10
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 557521 |
Immunogen | Mouse myeloma cell line NS0-derived transfected with recombinant human NPY2R Met1-Val381 Accession # P49146 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
Protein Aliases: Neuropeptide Y receptor type 2; neuropeptide Y receptor type 2 neuropeptide Y receptor Y2 NPY2-R NPY-Y2 receptor Y2 receptor; NPY-Y2 receptor; NPY2-R; Y2 receptor
Gene Aliases: NPY2-R; NPY2R
UniProt ID: (Human) P49146
Entrez Gene ID: (Human) 4887
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.