Invitrogen
Manufacturer Code:PA526477
Catalog # PIPA526477
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 19-45 amino acids from the N-terminal region of human NPY2R |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neuropeptide Y receptor type 2; NPY-Y2 receptor; NPY-Y2 receptor NPY2-R Y2 receptor neuropeptide Y receptor type 2; NPY2-R; Y2 receptor
Gene Aliases: NPY2-R; NPY2R
UniProt ID: (Human) P49146, (Mouse) P97295
Entrez Gene ID: (Human) 4887, (Mouse) 18167
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.