R&D Systems
Manufacturer Code:FAB6400R100UG
Catalog # FAB6400R100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 556153 |
Immunogen | NS0 mouse myeloma cell line transfected with human NPY1R Met1-Ile384 Accession # P25929 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: Neuropeptide Y receptor type 1; neuropeptide Y receptor type 1 neuropeptide Y receptor Y1 NPY1-R NPYR NPYY1; NPY1-R
Gene Aliases: NPY1-R; NPY1R; NPYR; NPYY1
UniProt ID: (Human) P25929
Entrez Gene ID: (Human) 4886
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.