Invitrogen
Manufacturer Code:PA576612
Catalog # PIPA576612
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human NPY1R. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: neuropeptide Y receptor subtype; Neuropeptide Y receptor type 1; Neuropeptide Y receptor type 1 NPY1-R NPY1R NPYR NPYY1; NPY-1 receptor; NPY1-R
Gene Aliases: NPY1-R; NPY1R; NPYR; NPYY1; Y1-R
UniProt ID: (Human) P25929, (Mouse) Q04573
Entrez Gene ID: (Human) 4886, (Mouse) 18166
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.