Invitrogen
Manufacturer Code:PA533989
Catalog # PIPA533989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 18 amino acid peptide from 3rd cytoplasmic domain of human NPY1R. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neuropeptide Y receptor type 1; NPY1-R; NPY1R NPYY1 NPY-Y1 receptor Neuropeptide Y receptor type 1 PY receptor Y1 Neuropeptide Y receptor Y1 Neuropeptide Y-Y1 receptor Neuropeptide Y1 receptor NPY1-R NPYR
Gene Aliases: NPY1-R; NPY1R; NPYR; NPYY1
UniProt ID: (Human) P25929
Entrez Gene ID: (Human) 4886
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.