Abnova
Manufacturer Code:H00004886B01P
Catalog # 89-002-580
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | NPY1R (AAH71720.1 1 a.a. ~ 384 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human NPY1R protein.
For Research Use Only
Protein Aliases: Neuropeptide Y receptor type 1; NPY1-R; NPYR
Gene Aliases: NPY1-R; NPY1R; NPYR; NPYY1
UniProt ID: (Human) P25929
Entrez Gene ID: (Human) 4886
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.