Invitrogen
Manufacturer Code:PA590205
Catalog # PIPA590205
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-97 of human NPY |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-flanking peptide of NPY; CPON; Neuropeptide tyrosine; Neuropeptide Y; NPY; prepro-neuropeptide Y; Pro-neuropeptide Y
Gene Aliases: NPY; PYY4
UniProt ID: (Human) P01303
Entrez Gene ID: (Human) 4852
Molecular Function:
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.