Invitrogen
Manufacturer Code:PA519568
Catalog # PIPA519568
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from within residues 1 to the C-terminus of Pig Neuropeptide Y. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-flanking peptide of NPY; CPON; involved in appetite regulation; Neuropeptide tyrosine; Neuropeptide Y; NPY; prepro-neuropeptide Y; Pro-neuropeptide Y; PYY4 OTTHUMP00000201947
Gene Aliases: 0710005A05Rik; NPY; NPY02; PYY4; RATNPY; RATNPY02
UniProt ID: (Human) P01303, (Mouse) P57774, (Rat) P07808
Entrez Gene ID: (Human) 4852, (Pig) 397304, (Mouse) 109648, (Rat) 24604
Molecular Function:
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.