Invitrogen
Manufacturer Code:PA130932
Catalog # PIPA130932
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Guinea pig |
Class | Polyclonal |
Type | Antibody |
Immunogen | SDLLMRESTENAPRTR Corresponding to residues 76 91 of rat preproneuropeptide Y |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Guinea Pig Polyclonal Antibody
For Research Use Only
Protein Aliases: C-flanking peptide of NPY; CPON; Neuropeptide tyrosine; Neuropeptide Y; NPY; Pro-neuropeptide Y
Gene Aliases: Npy; NPY02; RATNPY; RATNPY02
UniProt ID: (Rat) P07808
Entrez Gene ID: (Rat) 24604
Molecular Function: neuropeptide peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.