Invitrogen
Manufacturer Code:PA5111466
Catalog # PIPA5111466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human NPTX2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: apexin; NARP Neuronal pentraxin II Neuronal pentraxin-2 NP2 NP-II Nptx2; neuronal activity-regulated pentaxin; Neuronal pentraxin II; Neuronal pentraxin-2; NP-II; NP2
Gene Aliases: NARP; NP-II; NP2; NPTX2
UniProt ID: (Human) P47972
Entrez Gene ID: (Human) 4885
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.