Invitrogen
Manufacturer Code:PA5100295
Catalog # PIPA5100295
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NPTX2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: apexin; apexin Narp neuro activity regulated petaxin neuronal activity-regulated pentaxin neuronal activity-regulated pentraxin neuronal pentraxin 2 neuronal pentraxin II Neuronal pentraxin-2 np2 NP-II NPTX2; neuro activity regulated petaxin; neuronal activity-regulated pentaxin; neuronal activity-regulated pentraxin; neuronal pentraxin 2; Neuronal pentraxin II; Neuronal pentraxin-2; NP-II; NP2
Gene Aliases: NARP; NP-II; NP2; NPTX2
UniProt ID: (Human) P47972, (Mouse) O70340
Entrez Gene ID: (Human) 4885, (Rat) 288475, (Mouse) 53324
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.