Invitrogen
Manufacturer Code:PA525954
Catalog # PIPA525954
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 322-350 amino acids from human NPTX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neuronal pentraxin I; Neuronal pentraxin-1; NP-I; NP1; NP1 NP-I neuronal pentraxin-1
Gene Aliases: D11Bwg1004e; NP1; NPTX1
UniProt ID: (Human) Q15818, (Mouse) Q62443
Entrez Gene ID: (Human) 4884, (Mouse) 18164
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.