Invitrogen
Manufacturer Code:PA5114148
Catalog # PIPA5114148
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Immunogen sequence: EGKGGPKNDT EERAKIESAL TSLHQRISEL EKGQKDNRPG DKFQLTFPLR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 47 kDa taipoxin-binding protein D11Bwg1004e neuronal pentraxin 1 Neuronal pentraxin I Neuronal pentraxin-1 NP1 NP-I Nptx1; Neuronal pentraxin I; Neuronal pentraxin-1; NP-I; NP1
Gene Aliases: D11Bwg1004e; Np1; Nptx1
UniProt ID: (Mouse) Q62443
Entrez Gene ID: (Mouse) 18164
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.