Proteintech
Manufacturer Code:206561AP150UL
Catalog # 50-173-3017
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 47 kDa taipoxin-binding protein; neuronal pentraxin 1; Neuronal pentraxin 1 neuronal pentraxin I NPTX1; Neuronal pentraxin I; Neuronal pentraxin-1; NP-I; NP1
Gene Aliases: D11Bwg1004e; NP1; NPTX1
UniProt ID: (Human) Q15818, (Mouse) Q62443, (Rat) P47971
Entrez Gene ID: (Human) 4884, (Mouse) 18164, (Rat) 266777
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.