Proteintech
Manufacturer Code:207511AP150UL
Catalog # 50-173-3064
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NPT1 Fusion Protein Ag12819 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na(+)/PI cotransporter 1; Na(+)/PI cotransporter 1 Na/Pi 4 NAPI 1 SLC17A1; Na/Pi-4; Renal Na(+)-dependent phosphate cotransporter 1; Renal sodium-dependent phosphate transport protein 1; Renal sodium-phosphate transport protein 1; sodium phosphate transporter; Sodium-dependent phosphate transport protein 1; sodium-dependent phosphate/anion cotransporter; Sodium/phosphate cotransporter 1; sodium/phosphate type I cotransporter; solute carrier family 17 (organic anion transporter), member 1; solute carrier family 17 (sodium phosphate), member 1; solute carrier family 17 (sodium/hydrogen exchanger), member 1; solute carrier family 17 (vesicular glutamate transporter), member 1; Solute carrier family 17 member 1; solute carrier family 17 vesicular glutamate transporter), member 1
Gene Aliases: NAPI-1; Napi1; NPT-1; NPT1; SLC17A1
UniProt ID: (Human) Q14916, (Mouse) Q61983
Entrez Gene ID: (Human) 6568, (Rat) 171080, (Mouse) 20504
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.