Invitrogen
Manufacturer Code:PA524177
Catalog # PIPA524177
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 327-356 amino acids from the C-terminal region of human NPSR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ASRT2 GPR154 GPRA NPSR PGR14 VRR1 G protein-coupled receptor 154 G protein-coupled receptor for asthma susceptibility G-protein coupled receptor 154 G-protein coupled receptor PGR14 G-protein coupled receptor for asthma susceptibility neuropeptide S receptor vasopressin receptor-related r; G protein-coupled receptor 154; G protein-coupled receptor PGR14; G-protein coupled receptor 154; G-protein coupled receptor for asthma susceptibility; G-protein coupled receptor PGR14; Neuropeptide S receptor; vasopressin receptor-related receptor 1; vasotocin receptor
Gene Aliases: 9330128H10Rik; ASRT2; GPR154; GPRA; MVTR; NPSR; NPSR1; PGR14; VRR1
UniProt ID: (Human) Q6W5P4, (Mouse) Q8BZP8
Entrez Gene ID: (Human) 387129, (Mouse) 319239
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.