Invitrogen
Manufacturer Code:PA567690
Catalog # PIPA567690
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human NPRL2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810446G01Rik; G21 protein; GATOR complex protein NPRL2; Gene 21 protein; homologous to yeast nitrogen permease (candidate tumor suppressor); nitrogen permase homolog; Nitrogen permease regulator 2-like protein; NPR2-like protein; Tumor suppressor candidate 4; TUSC4 NPRL2 tumor suppressor candidate 4 NPR2L 2810446G01Rik homologous to yeast nitrogen permease (candidate tumor suppressor)
Gene Aliases: 2810446G01Rik; G21; NPR2; NPR2L; NPRL2; TUSC4
UniProt ID: (Human) A8K831, (Mouse) Q9WUE4
Entrez Gene ID: (Human) 10641, (Mouse) 56032, (Rat) 363138
Molecular Function: nuclease nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.