Invitrogen
Manufacturer Code:PA5113933
Catalog # PIPA5113933
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal region of human TUSC4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810446G01Rik; 2810446G01Rik G21 G21 protein GATOR complex protein NPRL2 Gene 21 protein homologous to yeast nitrogen permease (candidate tumor suppressor) nitrogen permase homolog Nitrogen permease regulator 2-like protein nitrogen permease regulator-like 2 nitrogen permease regulator-like 2 (S. cerevisiae) NPR 2L NPR L2 NPR like 2 NPR2 NPR2 like NPR2L NPR2-like protein NPR2-like GATOR1 complex subunit NPRL 2 Nprl2 Tumor suppressor candidate 4 TUSC 4 TUSC 4 protein TUSC4 TUSC4 protein; G21 protein; GATOR complex protein NPRL2; Gene 21 protein; homologous to yeast nitrogen permease (candidate tumor suppressor); Nitrogen permease regulator 2-like protein; NPR2-like protein; Tumor suppressor candidate 4
Gene Aliases: NPR2; NPR2L; NPRL2; TUSC4
UniProt ID: (Human) A8K831
Entrez Gene ID: (Human) 10641
Molecular Function: nuclease nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.