OriGene
Manufacturer Code:TA501046
Catalog # 50-167-2189
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4A1 |
Immunogen | Full length human recombit protein of human NPR3 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ANP-C; ANP-C ANPR-C ANPRC C5orf23 GUCY2B NPR-C NPRC; Atrial natriuretic peptide clearance receptor; Atrial natriuretic peptide receptor 3; Atrial natriuretic peptide receptor type C; atrionatriuretic peptide receptor C; guanylate cyclase C; natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptide receptor C)
Gene Aliases: ANP-C; ANPR-C; ANPRC; C5orf23; GUCY2B; NPR-C; NPR3; NPRC
UniProt ID: (Human) P17342
Entrez Gene ID: (Human) 4883
Molecular Function: adenylate cyclase cyclase guanylate cyclase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.