Invitrogen
Manufacturer Code:PA599836
Catalog # PIPA599836
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NPR2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMDM ANPb ANP-B ANPRB ANPR-B atrial natriuretic peptide B-type receptor atrial natriuretic peptide receptor 2 atrial natriuretic peptide receptor B Atrial natriuretic peptide receptor type B cn ECDM GC-B GC-B2 GC-B3 guanylate cyclase 2B Guanylate cyclase B guanylyl cyclase-B GUC2B GUCY2B mNPR-B natriuretic peptide receptor 2 natriuretic peptide receptor B type natriuretic peptide receptor B/guanylate cyclase B (atrionatriuretic peptide receptor B) natriuretic peptide receptor-B Nppb receptor NPR2 NPRB NPR-B NPRBi pwe SNSK; ANP-B; ANPR-B; ANPRB; atrial natriuretic peptide B-type receptor; Atrial natriuretic peptide receptor 2; atrial natriuretic peptide receptor B; Atrial natriuretic peptide receptor type B; GC-B; guanylate cyclase 2B; Guanylate cyclase B; guanylyl cyclase-B; natriuretic peptide receptor B type; natriuretic peptide receptor B/guanylate cyclase B (atrionatriuretic peptide receptor B); natriuretic peptide receptor-B; Nppb receptor; NPR-B
Gene Aliases: AMDM; ANPb; ANPRB; cn; ECDM; GC-B; GC-B2; GC-B3; GUC2B; GUCY2B; mNPR-B; NPR-B; NPR2; NPRB; NPRBi; pwe; SNSK
UniProt ID: (Human) P20594, (Rat) P16067, (Mouse) Q6VVW5
Entrez Gene ID: (Human) 4882, (Rat) 116564, (Mouse) 230103
Molecular Function: adenylate cyclase cyclase guanylate cyclase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.