Abnova
Manufacturer Code:H00004882A01
Catalog # 89-105-022
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 131 a.a. &bsim 230 a.a) partial recombinant protein with GST tag.,NPR2 (AAH34917 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant NPR2.
For Research Use Only
Protein Aliases: AMDM; ANP-B; ANPR-B; ANPRB; atrial natriuretic peptide B-type receptor; Atrial natriuretic peptide receptor 2; Atrial natriuretic peptide receptor type B; GC-B; guanylate cyclase 2B; Guanylate cyclase B; GUC2B; GUCY2B; natriuretic peptide receptor B/guanylate cyclase B (atrionatriuretic peptide receptor B); NPR-B; NPRB; NPRBi
Gene Aliases: AMDM; ANPb; ANPRB; ECDM; GUC2B; GUCY2B; NPR2; NPRB; NPRBi; SNSK
UniProt ID: (Human) P20594
Entrez Gene ID: (Human) 4882
Molecular Function: adenylate cyclase cyclase guanylate cyclase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.